ADVH49B836EA AChemBlock

ethyl 5-bromo-2-iodobenzoate

  • Cas Number : 450412-27-8
  • Molecular Weight : 354.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-5-bromo-2-iodobenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89409
Availability details for
Quantity
  • Availability
  • Add to Cart