ADVH49B836EB AChemBlock

4-amino-5-chloro-2-methoxy-3-nitrobenzoic acid

  • Cas Number : 457947-61-4
  • Molecular Weight : 246.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-amino-5-chloro-2-methoxy-3-nitrobenzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89410
Availability details for
Quantity
  • Availability
  • Add to Cart