ADVH49B836EE AChemBlock

6-(4-methoxyphenyl)pyridazin-3-amine

  • Cas Number : 4776-87-8
  • Molecular Weight : 201.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-4-methoxyphenyl-pyridazin-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89413
Availability details for
Quantity
  • Availability
  • Add to Cart