ADVH49B836F1 AChemBlock

3-(4-methyl-2,5-dioxo-2,5-dihydrofuran-3-yl)propanoic acid

  • Cas Number : 487-66-1
  • Molecular Weight : 184.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-4-methyl-2-5-dioxo-2-5-dihydrofuran-3-yl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89416
Availability details for
Quantity
  • Availability
  • Add to Cart