ADVH49B836F3 AChemBlock

tert-butyl 2-iodoacetate

  • Cas Number : 49827-15-8
  • Molecular Weight : 242.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-iodoacetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89418
Availability details for
Quantity
  • Availability
  • Add to Cart