ADVH49B836F4 AChemBlock

methyl 4-((tert-butoxycarbonyl)amino)-1-methyl-1H-imidazole-2-carboxylate

  • Cas Number : 500701-36-0
  • Molecular Weight : 255.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-tert-butoxycarbonyl-amino-1-methyl-1h-imidazole-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89419
Availability details for
Quantity
  • Availability
  • Add to Cart