ADVH49B836FF AChemBlock

N,N-bis(2-hydroxypropyl)nitrous amide

  • Cas Number : 53609-64-6
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-n-bis-2-hydroxypropyl-nitrous-amide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89430
Availability details for
Quantity
  • Availability
  • Add to Cart