ADVH49B83702 AChemBlock

5-(benzyloxy)-5-oxopentanoic acid

  • Cas Number : 54322-10-0
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-benzyloxy-5-oxopentanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89433
Availability details for
Quantity
  • Availability
  • Add to Cart