ADVH49B83704 AChemBlock

7-hydroxychroman-2-one

  • Cas Number : 5631-67-4
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-hydroxychroman-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89435
Availability details for
Quantity
  • Availability
  • Add to Cart