ADVH49B83707 AChemBlock

2-(3-chlorophenyl)acetamide

  • Cas Number : 58357-84-9
  • Molecular Weight : 169.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-3-chlorophenyl-acetamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89438
Availability details for
Quantity
  • Availability
  • Add to Cart