ADVH49B83708 AChemBlock

3-benzyl-7-chloro-2-isobutylquinazolin-4(3H)-one

  • Cas Number : 587881-23-0
  • Molecular Weight : 326.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-benzyl-7-chloro-2-isobutylquinazolin-4-3h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89439
Availability details for
Quantity
  • Availability
  • Add to Cart