ADVH49B8370A AChemBlock

4-(bromomethyl)benzamide

  • Cas Number : 58914-40-2
  • Molecular Weight : 214.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromomethyl-benzamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89441
Availability details for
Quantity
  • Availability
  • Add to Cart