ADVH49B8370D AChemBlock

2,3-dichloro-4-nitrophenol

  • Cas Number : 59384-57-5
  • Molecular Weight : 207.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-3-dichloro-4-nitrophenol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89444
Availability details for
Quantity
  • Availability
  • Add to Cart