ADVH49B8370F AChemBlock

(1-phenylpropan-2-yl)glycine hydrochloride

  • Cas Number : 60342-93-0
  • Molecular Weight : 229.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-phenylpropan-2-yl-glycine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89446
Availability details for
Quantity
  • Availability
  • Add to Cart