ADVH49B83713 AChemBlock

ethyl 7-hydroxy-2-oxo-2H-chromene-3-carboxylate

  • Cas Number : 6093-71-6
  • Molecular Weight : 234.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-7-hydroxy-2-oxo-2h-chromene-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89450
Availability details for
Quantity
  • Availability
  • Add to Cart