ADVH49B83716 AChemBlock

4-(4-chlorobenzyl)benzene-1,3-diol

  • Cas Number : 6280-43-9
  • Molecular Weight : 234.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-4-chlorobenzyl-benzene-1-3-diol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89453
Availability details for
Quantity
  • Availability
  • Add to Cart