ADVH49B8371D AChemBlock

ethyl 2-amino-5,5-dimethyl-4,7-dihydro-5H-thieno[2,3-c]pyran-3-carboxylate

  • Cas Number : 64792-51-4
  • Molecular Weight : 255.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-amino-5-5-dimethyl-4-7-dihydro-5h-thieno-2-3-c-pyran-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89460
Availability details for
Quantity
  • Availability
  • Add to Cart