ADVH49B8371E AChemBlock

5-(benzylthio)-N-methyl-1,3,4-thiadiazol-2-amine

  • Cas Number : 65373-29-7
  • Molecular Weight : 237.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-benzylthio-n-methyl-1-3-4-thiadiazol-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89461
Availability details for
Quantity
  • Availability
  • Add to Cart