ADVH49B83720 AChemBlock

6-methylbenzo[d]isoxazol-3(2H)-one

  • Cas Number : 66571-26-4
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-methylbenzo-d-isoxazol-3-2h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89463
Availability details for
Quantity
  • Availability
  • Add to Cart