ADVH49B83724 AChemBlock

5-(5-acetyl-6-methyl-2-oxo-1-(3-(trifluoromethyl)phenyl)-1,2,3,4-tetrahydropyrimidin-4-yl)picolinonitrile

  • Cas Number : 675103-35-2
  • Molecular Weight : 400.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-5-acetyl-6-methyl-2-oxo-1-3-trifluoromethyl-phenyl-1-2-3-4-tetrahydropyrimidin-4-yl-picolinonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89467
Availability details for
Quantity
  • Availability
  • Add to Cart