ADVH49B83725 AChemBlock

6-bromobenzo[d]isothiazole-3-carboxylic acid

  • Cas Number : 677304-75-5
  • Molecular Weight : 258.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromobenzo-d-isothiazole-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89468
Availability details for
Quantity
  • Availability
  • Add to Cart