ADVH49B83728 AChemBlock

2-methyl-1H-imidazo[4,5-b]pyridine

  • Cas Number : 68175-07-5
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-1h-imidazo-4-5-b-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89471
Availability details for
Quantity
  • Availability
  • Add to Cart