ADVH49B83729 AChemBlock

dimethyl 2-(5-nitropyridin-2-yl)malonate

  • Cas Number : 68719-87-9
  • Molecular Weight : 254.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/dimethyl-2-5-nitropyridin-2-yl-malonate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89472
Availability details for
Quantity
  • Availability
  • Add to Cart