ADVH49B8372A AChemBlock

4-(1H-tetrazol-1-yl)piperidine

  • Cas Number : 690261-90-6
  • Molecular Weight : 153.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-1h-tetrazol-1-yl-piperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89473
Availability details for
Quantity
  • Availability
  • Add to Cart