ADVH49B8372B AChemBlock

diethyl 2-hydroxypentanedioate

  • Cas Number : 69134-53-8
  • Molecular Weight : 204.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/diethyl-2-hydroxypentanedioate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89474
Availability details for
Quantity
  • Availability
  • Add to Cart