ADVH49B8372C AChemBlock

tetrahydro-2H-pyran-2-ol

  • Cas Number : 694-54-2
  • Molecular Weight : 102.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tetrahydro-2h-pyran-2-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89475
Availability details for
Quantity
  • Availability
  • Add to Cart