ADVH49B83731 AChemBlock

1-(6-aminohexyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate

  • Cas Number : 731862-92-3
  • Molecular Weight : 310.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-6-aminohexyl-1h-pyrrole-2-5-dione-2-2-2-trifluoroacetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89480
Availability details for
Quantity
  • Availability
  • Add to Cart