ADVH49B83736 AChemBlock

1-(4-bromo-3-fluorophenyl)cyclopropane-1-carboxylic acid

  • Cas Number : 749269-74-7
  • Molecular Weight : 259.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-4-bromo-3-fluorophenyl-cyclopropane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89485
Availability details for
Quantity
  • Availability
  • Add to Cart