ADVH49B83739 AChemBlock

2,2'-(propane-1,3-diyldisulfinyl)bis(ethan-1-ol)

  • Cas Number : 76305-83-4
  • Molecular Weight : 228.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2-propane-1-3-diyldisulfinyl-bis-ethan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89488
Availability details for
Quantity
  • Availability
  • Add to Cart