ADVH49B8373B AChemBlock

ethyl 4-fluoro-3-methoxybenzoate

  • Cas Number : 773136-55-3
  • Molecular Weight : 198.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-fluoro-3-methoxybenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89490
Availability details for
Quantity
  • Availability
  • Add to Cart