ADVH49B8373C AChemBlock

ethyl 2-fluoro-5-iodobenzoate

  • Cas Number : 773136-66-6
  • Molecular Weight : 294.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-fluoro-5-iodobenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89491
Availability details for
Quantity
  • Availability
  • Add to Cart