ADVH49B8373E AChemBlock

3-chloro-6-(pyridin-4-yl)pyridazine

  • Cas Number : 79472-17-6
  • Molecular Weight : 191.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-6-pyridin-4-yl-pyridazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89493
Availability details for
Quantity
  • Availability
  • Add to Cart