ADVH49B8373F AChemBlock

2,6-dibromo-3-methoxypyridine

  • Cas Number : 79491-45-5
  • Molecular Weight : 266.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-6-dibromo-3-methoxypyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89494
Availability details for
Quantity
  • Availability
  • Add to Cart