ADVH49B83741 AChemBlock

2-(2-methoxyphenyl)propanoic acid

  • Cas Number : 81616-80-0
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2-methoxyphenyl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89496
Availability details for
Quantity
  • Availability
  • Add to Cart