ADVH49B83744 AChemBlock

6-methoxy-1-(3-sulfopropyl)quinolin-1-ium

  • Cas Number : 83907-40-8
  • Molecular Weight : 282.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-methoxy-1-3-sulfopropyl-quinolin-1-ium.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89499
Availability details for
Quantity
  • Availability
  • Add to Cart