ADVH49B83745 AChemBlock

5-(piperazin-1-ylsulfonyl)isoquinoline

  • Cas Number : 84468-24-6
  • Molecular Weight : 277.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-piperazin-1-ylsulfonyl-isoquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89500
Availability details for
Quantity
  • Availability
  • Add to Cart