ADVH49B83748 AChemBlock

2,2-dimethyl-7-nitro-2H-benzo[b][1,4]oxazin-3(4H)-one

  • Cas Number : 85160-83-4
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2-dimethyl-7-nitro-2h-benzo-b-1-4-oxazin-3-4h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89503
Availability details for
Quantity
  • Availability
  • Add to Cart