ADVH49B8374A AChemBlock

5-((1,3-dioxoisoindolin-2-yl)methyl)thiophene-2-sulfonamide

  • Cas Number : 859491-30-8
  • Molecular Weight : 322.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-1-3-dioxoisoindolin-2-yl-methyl-thiophene-2-sulfonamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89505
Availability details for
Quantity
  • Availability
  • Add to Cart