ADVH49B8374B AChemBlock

1-bromo-3-iodo-5-nitrobenzene

  • Cas Number : 861601-15-2
  • Molecular Weight : 327.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-bromo-3-iodo-5-nitrobenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89506
Availability details for
Quantity
  • Availability
  • Add to Cart