ADVH49B8374C AChemBlock

5-bromo-2-ethyl-4-methylthiazole

  • Cas Number : 863190-90-3
  • Molecular Weight : 206.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-ethyl-4-methylthiazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89507
Availability details for
Quantity
  • Availability
  • Add to Cart