ADVH49B8374D AChemBlock

methyl 4-bromo-1-methyl-1H-imidazole-2-carboxylate

  • Cas Number : 864076-05-1
  • Molecular Weight : 219.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-bromo-1-methyl-1h-imidazole-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89508
Availability details for
Quantity
  • Availability
  • Add to Cart