ADVH49B8374E AChemBlock

2-(2-chlorothiazol-5-yl)acetonitrile

  • Cas Number : 865660-15-7
  • Molecular Weight : 158.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2-chlorothiazol-5-yl-acetonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89509
Availability details for
Quantity
  • Availability
  • Add to Cart