ADVH49B83750 AChemBlock

4-(1H-imidazol-1-yl)-3-methoxybenzaldehyde

  • Cas Number : 870837-70-0
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-1h-imidazol-1-yl-3-methoxybenzaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89511
Availability details for
Quantity
  • Availability
  • Add to Cart