ADVH49B83751 AChemBlock

2-(bromomethyl)benzamide

  • Cas Number : 872414-52-3
  • Molecular Weight : 214.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-bromomethyl-benzamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89512
Availability details for
Quantity
  • Availability
  • Add to Cart