ADVH49B83752 AChemBlock

ethyl 5-sulfamoylfuran-2-carboxylate

  • Cas Number : 87299-64-7
  • Molecular Weight : 219.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-5-sulfamoylfuran-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89513
Availability details for
Quantity
  • Availability
  • Add to Cart