ADVH49B83754 AChemBlock

N-(3-bromo-4-methylphenyl)-3-(trifluoromethyl)benzamide

  • Cas Number : 876322-59-7
  • Molecular Weight : 358.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-3-bromo-4-methylphenyl-3-trifluoromethyl-benzamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89515
Availability details for
Quantity
  • Availability
  • Add to Cart