ADVH49B83756 AChemBlock

2-chloro-3,4-dihydroxybenzoic acid

  • Cas Number : 87932-50-1
  • Molecular Weight : 188.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-3-4-dihydroxybenzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89517
Availability details for
Quantity
  • Availability
  • Add to Cart