ADVH49B83758 AChemBlock

2-((1H-imidazol-1-yl)methyl)benzoic acid

  • Cas Number : 883543-82-6
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-1h-imidazol-1-yl-methyl-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89519
Availability details for
Quantity
  • Availability
  • Add to Cart