ADVH49B83759 AChemBlock

N-methyl-1-(5-methyl-1H-benzo[d]imidazol-2-yl)methanamine

  • Cas Number : 887571-32-6
  • Molecular Weight : 175.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-methyl-1-5-methyl-1h-benzo-d-imidazol-2-yl-methanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89520
Availability details for
Quantity
  • Availability
  • Add to Cart