ADVH49B8375C AChemBlock

4-formyl-3-nitrobenzonitrile

  • Cas Number : 90178-78-2
  • Molecular Weight : 176.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-formyl-3-nitrobenzonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89523
Availability details for
Quantity
  • Availability
  • Add to Cart